EMAIL THIS PAGE TO A FRIEND
Journal of Neuroendocrinology
Direct regulation of gonadotrophin-releasing hormone (GnRH) transcription by RF-amide-related peptide-3 and kisspeptin in a novel GnRH-secreting cell line, mHypoA-GnRH/GFP.
PMID 25283492
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1916
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
C12H12Cl2N2O4