EMAIL THIS PAGE TO A FRIEND
Nature communications
Stabilization and targeting of INO80 to replication forks by BAP1 during normal DNA synthesis.
PMID 25283999
Abstract
The INO80 chromatin-remodelling complex has been implicated in DNA replication during stress in yeast. However, its role in normal DNA replication and its underlying mechanisms remain unclear. Here, we show that INO80 binds to replication forks and promotes fork progression in human cells under unperturbed, normal conditions. We find that Ino80, which encodes the catalytic ATPase of INO80, is essential for mouse embryonic DNA replication and development. Ino80 is recruited to replication forks through interaction with ubiquitinated H2A--aided by BRCA1-associated protein-1 (BAP1), a tumour suppressor and nuclear de-ubiquitinating enzyme that also functions to stabilize Ino80. Importantly, Ino80 is downregulated in BAP1-defective cancer cells due to the lack of an Ino80 stabilization mechanism via BAP1. Our results establish a role for INO80 in normal DNA replication and uncover a mechanism by which this remodeler is targeted to replication forks, suggesting a molecular basis for the tumour-suppressing function of BAP1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50553
α-Tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Ca3O8P2
SAB1406654
Anti-BAP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
M5691 Anti-MAGI-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA031853
Anti-MAGI1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA026803
Anti-RNF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
16483
β-tri-Calcium phosphate, puriss. p.a., ≥95% β-phase basis (sintered powder)
Ca3O8P2
SAB1404687
Monoclonal Anti-MAGI1 antibody produced in mouse, clone 3B4, purified immunoglobulin, buffered aqueous solution
WH0009223M3
Monoclonal Anti-MAGI1 antibody produced in mouse, clone 7B4, purified immunoglobulin, buffered aqueous solution
WH0006045M1
Monoclonal Anti-RNF2 antibody produced in mouse, clone 6C2, purified immunoglobulin, buffered aqueous solution
PLA0248
Rabbit anti-BAP1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
AV34289
Anti-RNF2 (AB1) antibody produced in rabbit, affinity isolated antibody
AV34290
Anti-RNF2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1404320
Monoclonal Anti-RNF2 antibody produced in mouse, clone 2B6, purified immunoglobulin, buffered aqueous solution