EMAIL THIS PAGE TO A FRIEND
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians
Monitoring neonatal fungal infection with metabolomics.
PMID 25284175
Abstract
The objective of our study was to evaluate the capability of the metabolomics approach to identify the variations of urine metabolites over time related to the neonatal fungal septic condition. The study population included a clinical case of a preterm neonate with invasive fungal infection and 13 healthy preterm controls. This study showed a unique urine metabolic profile of the patient affected by fungal sepsis compared to urine of controls and it was also possible to evaluate the efficacy of therapy in improving patient health.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H50004
4-Hydroxyphenylacetic acid, 98%
C8H8O3
A7377
D-Alanine, ≥98% (HPLC)
C3H7NO2
T8250
D-Threonine, ≥98% (TLC)
C4H9NO3
240176
DL-Malic acid, ReagentPlus®, ≥99%
C4H6O5
M0875
DL-Malic acid, ≥98% (capillary GC)
C4H6O5
94916
DL-Malic acid, analytical standard
C4H6O5
27606
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
C4H6O5
W265501
DL-Malic acid, ≥99%
C4H6O5
69673
Homovanillic acid, analytical standard
C9H10O4
S4500
L-Serine, ReagentPlus®, ≥99% (HPLC)
C3H7NO3
RES0908S-A7
L-Serine, PharmaGrade, Ajinomoto, EP, USP, JP, Manufactured under appropriate controls for use in pharma or biopharmaceutical production, suitable for cell culture
C3H7NO3
S4311
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C3H7NO3
84959
L-Serine, BioUltra, ≥99.5% (NT)
C3H7NO3
54763
L-Serine, certified reference material, TraceCERT®
C3H7NO3
PHR1103
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO3
M8304
Malic acid, meets USP/NF testing specifications
C4H6O5
PHR1273
Malic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C4H6O5
1374601
Malic acid, United States Pharmacopeia (USP) Reference Standard
C4H6O5
04621
Oxalate standard for IC, 1.000 g/L in H2O, analytical standard
C2H2O4
68487
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
C2H2O4
X3375
Xylitol, ≥99%
C5H12O5
W507930
Xylitol
C5H12O5
47844
Xylitol, analytical standard
C5H12O5
PHR1280
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
C5H12O5
X0080000
Xylitol, European Pharmacopoeia (EP) Reference Standard
C5H12O5
1720600
Xylitol, United States Pharmacopeia (USP) Reference Standard
C5H12O5