EMAIL THIS PAGE TO A FRIEND
Indian journal of pharmaceutical sciences
Olea europaea Linn. Fruit Pulp Extract Protects against Carbon Tetrachloride-induced Hepatic Damage in Mice.
PMID 25284924
Abstract
The present study we investigated the hepatoprotective effects of Olea europaea fruit pulp extract against carbon tetrachloride-induced hepatic damage in experimental mice. Further we explored the antioxidant potential of the extract to substantiate the hepatoprotective properties. Biochemical parameters were analyzed in the serum of experimental mice using respective diagnostic kits. Antioxidant activities were measured following alkyl and hydroxyl radical scavenging assays. Compared with control groups, administration of the extract to carbon tetrachloride-treated mice significantly reduced the elevated serum levels of alanine aminotransferase, aspartate aminotransferase, and alkaline phosphatase. The carbon tetrachloride-treated morphological changes in hepatocyte architecture were also reversed by extract pretreatment. Further, the carbon tetrachloride-treated increased serum cholesterol levels such as triglyceride and low density/very low-density lipoprotein in the liver were reversed in acute and chronic carbon tetrachloride-treated mice. The extract was also found to significantly increase the serum level of high-density lipoproteins in carbon tetrachloride-treated mice. Furthermore, the extract showed significant in vitro antioxidant actions by scavenging the alkyl and hydroxyl free radicals, substantiating its use in hepatoprotection. The concentration of the extract necessary for 50% inhibition of alkyl and hydroxyl radicals was 72.41 and 52.24 μg/ml, respectively. In conclusion, data from our study suggest that Olea europaea fruit pulp extract could prevent carbon tetrachloride-treated acute and chronic liver degeneration and attenuated the lipid levels elevated by carbon tetrachloride. The hepatoprotective activity exhibited by Olea europaea extract might possibly be through its antioxidant defense mechanisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92688
5,5-Dimethyl-1-pyrroline N-oxide, for ESR-spectroscopy
C6H11NO