EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Radical scavenging and antioxidant effects of Matricaria chamomilla polyphenolic-polysaccharide conjugates.
PMID 25285848
Abstract
Matricaria chamomilla L. (MC), a member of the Asteraceae family, is one of the oldest medicinal plants, widely used worldwide for a variety of healing applications. Its recommendations, derived from both traditional and modern medicine, include numerous disorders such as inflammation, ulcers, wounds, gastrointestinal disorders, stomach ache, pharyngitis, rheumatic pain, as well as the other ailments. This work is focused on another aspect of the biological activity of chamomile polyphenolic-polysaccharide conjugates--their antioxidant properties in the protection of blood plasma components against in vitro oxidative stress. Measurements of DPPH and ABTS radical scavenging indicated considerable anti-free radical action of MC. Pre-incubation of blood plasma with MC considerably diminished the extent of ONOO(-)-induced oxidative modifications such as protein carbonyl groups, SH groups, 3-nitrotyrosine, as well as the formation of lipid hydroperoxides. The analysis of the FRAP assay result shows a considerable increase of ferric reducing ability of blood plasma in the presence of MC. The results obtained in this study indicate that polyphenolic-polysaccharide conjugates isolated from M. chamomilla substances possess antioxidant properties. The M. chamomilla macromolecular glycoconjugates may be useful in the creation of new natural-based medications or dietary supplements, helpful in the prevention and treatment of oxidative stress-mediated disorders.