EMAIL THIS PAGE TO A FRIEND
Acta Crystallographica. Section F, Structural Biology Communications
Expression, crystallization and preliminary X-ray analysis of McbB, a multifunctional enzyme involved in ?-carboline skeleton biosynthesis.
PMID 25286949