EMAIL THIS PAGE TO A FRIEND
Hypertension (Dallas, Tex. : 1979)
Blood pressure in relation to environmental lead exposure in the national health and nutrition examination survey 2003 to 2010.
PMID 25287397
Abstract
In view of the declining environmental lead exposure in the United States, we analyzed the National Health and Nutrition Examination Survey (2003-2010) for association of blood pressure and hypertension with blood lead. The 12 725 participants included 21.1% blacks, 20.5% Hispanics, 58.4% whites, and 48.7% women. Blacks compared with non-Blacks had higher systolic and diastolic pressures (126.5 versus 123.9 and 71.9 versus 69.6 mm Hg) and higher hypertension prevalence (44.7 versus 36.8%). Blood lead was lower in whites than in non-whites (1.46 versus 1.57 μg/dL) and in women than in men (1.25 versus 1.80 μg/dL). In multivariable analyses of all participants, blood lead doubling was associated with higher (P≤0.0007) systolic and diastolic pressure (+0.76 mm Hg; 95% confidence interval, 0.38-1.13 and +0.43 mm Hg; 0.18-0.68), but not with the odds of hypertension (0.95; 0.90-1.01; P=0.11). Associations with blood lead were nonsignificant (P≥0.09) for systolic pressure in women and for diastolic pressure in non-whites. Among men, systolic pressure increased with blood lead (P≤0.060) with effect sizes associated with blood lead doubling ranging from +0.65 mm Hg in whites to +1.61 mm Hg in blacks. For systolic pressure, interactions of ethnicity and sex with blood lead were all significant (P≤0.019). In conclusion, small and inconsistent effect sizes in the associations of blood pressure with blood lead likely exclude current environmental lead exposure as a major hypertension cause in the United States.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
391352
Lead, powder, −100 mesh, 99.95% trace metals basis
Pb
209708
Lead, powder, −325 mesh, ≥99% trace metals basis
Pb
396117
Lead, shot, <2 mm, 99.9% trace metals basis
Pb
265888
Lead, wire, diam. 1.0 mm, 99.99% trace metals basis
Pb
695912
Lead, shot, 1-3 mm, 99.995% trace metals basis
Pb
GF04438919
Lead, foil, 50x50mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF21166321
Lead, foil, 100x100mm, thickness 3.0mm, as rolled, 99.95%
Pb
GF24521329
Lead, foil, 100x100mm, thickness 3.0mm, as rolled, 99.99+%
Pb
GF97211572
Lead, foil, 100x100mm, thickness 6.0mm, as rolled, 99.99+%
Pb
GF26692263
Lead, foil, 150x150mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF86984229
Lead, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF22298777
Lead, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF58005301
Lead, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF21793372
Lead, foil, 150x150mm, thickness 0.55mm, as rolled, 99.999%
Pb
GF96512597
Lead, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF74259793
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.5%
Pb
GF57430638
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Pb
GF60047760
Lead, foil, 250x250mm, thickness 3.0mm, as rolled, 99.95%
Pb
GF05521160
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF42741637
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Pb
GF60380940
Lead, foil, 25x25mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF85562994
Lead, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF74435519
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF10987757
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Pb
GF03985724
Lead, foil, 25x25mm, thickness 0.55mm, as rolled, 99.999%
Pb
GF73081852
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF68898188
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Pb
GF90025129
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.5%
Pb
GF55764041
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.95%
Pb
GF85349346
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Pb