EMAIL THIS PAGE TO A FRIEND
Pharmaceutical biology
Effect of parthenolide on growth and apoptosis regulatory genes of human cancer cell lines.
PMID 25289524
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0667
Parthenolide, ≥98% (HPLC)
C15H20O3