PloS one
Regulation of the tyrosine phosphorylation of Phospholipid Scramblase 1 in mast cells that are stimulated through the high-affinity IgE receptor.
PMID 25289695
Abstract
Engagement of high-affinity immunoglobulin E receptors (FcεRI) activates two signaling pathways in mast cells. The Lyn pathway leads to recruitment of Syk and to calcium mobilization whereas the Fyn pathway leads to phosphatidylinositol 3-kinase recruitment. Mapping the connections between both pathways remains an important task to be completed. We previously reported that Phospholipid Scramblase 1 (PLSCR1) is phosphorylated on tyrosine after cross-linking FcεRI on RBL-2H3 rat mast cells, amplifies mast cell degranulation, and is associated with both Lyn and Syk tyrosine kinases. Here, analysis of the pathway leading to PLSCR1 tyrosine phosphorylation reveals that it depends on the FcRγ chain. FcεRI aggregation in Fyn-deficient mouse bone marrow-derived mast cells (BMMC) induced a more robust increase in FcεRI-dependent tyrosine phosphorylation of PLSCR1 compared to wild-type cells, whereas PLSCR1 phosphorylation was abolished in Lyn-deficient BMMC. Lyn association with PLSCR1 was not altered in Fyn-deficient BMMC. PLSCR1 phosphorylation was also dependent on the kinase Syk and significantly, but partially, dependent on detectable calcium mobilization. Thus, the Lyn/Syk/calcium axis promotes PLSCR1 phosphorylation in multiple ways. Conversely, the Fyn-dependent pathway negatively regulates it. This study reveals a complex regulation for PLSCR1 tyrosine phosphorylation in FcεRI-activated mast cells and that PLSCR1 sits at a crossroads between Lyn and Fyn pathways.