EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Chiral liquid chromatography-tandem mass spectrometry assay to determine that dexpramipexole is not converted to pramipexole in vivo after administered in humans.
PMID 25289790