EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Sequential intravenous injection of anionic polymer and cationic lipoplex of siRNA could effectively deliver siRNA to the liver.
PMID 25290812
Abstract
In this study, we developed novel siRNA transfer method to the liver by sequential intravenous injection of anionic polymer and cationic liposome/cholesterol-modified siRNA complex (cationic lipoplex). When cationic lipoplex was intravenously injected into mice, the accumulation of siRNA was mainly observed in the lungs. In contrast, when cationic lipoplex was intravenously injected at 1 min after intravenous injection of poly-L-glutamic acid (PGA) or chondroitin sulfate C (CS), siRNA was accumulated in the liver. In terms of suppression of gene expression in vivo, apolipoprotein B (ApoB) mRNA in the liver and low-density-lipoprotein (LDL) and very low-density-lipoprotein (VLDL) cholesterol level in serum were reduced at 48 h after single sequential injection of PGA or CS plus cationic lipoplex of cholesterol-modified ApoB siRNA. Furthermore, sequential injections of PGA plus cationic lipoplex of cholesterol-modified luciferase siRNA could reduce luciferase activity in tumor xenografts bearing liver metastasis of human breast tumor MCF-7-Luc. From these findings, sequential injection of anionic polymer and cationic lipoplex of siRNA might produce a systemic vector of siRNA to the liver.