EMAIL THIS PAGE TO A FRIEND
Clinical and Experimental Immunology
Regulation of the alternative pathway of complement modulates injury and immunity in a chronic model of dextran sulphate sodium-induced colitis.
PMID 25293413