EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Antimicrobial activity, toxicity and selectivity index of two biflavonoids and a flavone isolated from Podocarpus henkelii (Podocarpaceae) leaves.
PMID 25293523
Abstract
Different parts of Podocarpus henkelii have been used in many cultures around the world to treat ailments such as cholera, stomach diseases, rheumatism, cancer, canine distemper in dogs and gall sickness in cattle. The aim of this study was to evaluate the biological activity and toxicity of isolated compounds from Podocarpus henkelii after an earlier study indicated a promising activity in crude extracts against viral pathogens of veterinary importance. The antibacterial and antifungal activity of two biflavonoids 7, 4', 7", 4"'-tetramethoxy amentoflavone (TMA), isoginkgetin (IGG) and podocarpus flavone-A (PFA) isolated from the leaves of Podocarpus henkelii were determined using a serial microplate dilution method with tetrazolium violet as growth indicator. The cytotoxicity of compounds TMA and IGG were determined on different cell types using a tetrazolium-based colorimetric cellular assay (MTT). The Ames test was used to determine their mutagenic activities. TMA had reasonable antifungal activity against Aspergillus fumigatus (MIC = 30 μg/ml). IGG had a wide spectrum of activity against four bacterial and two fungal pathogens with much higher selectivity index values obtained for A. fumigatus and Cryptococcus neoformans (SI > 30). PFA had a broad spectrum of activity against Enterococcus faecalis and Pseudomonas aeruginosa (SI > 15) and less activity against the two fungal pathogens. In both the cytotoxicity assays and Ames mutagenicity test using Salmonella typhimurium strains TA98 and TA100, TMA and IGG had no deleterious effect on the different cell types and did not induce mutations in the Ames test. Although the antimicrobial activities of the isolated compounds were not that exciting, the compounds had no cytotoxic activity at the highest concentration (1000 μg/ml) tested against all three cell lines. IGG was the most active against E. coli, S. aureus, A. fumigatus and C. neoformans, exhibiting both antibacterial and antifungal activity with good selectivity index values. PFA had a broad spectrum of activity against E. faecalis and P. aeruginosa. The two compounds isolated had low toxicity and no genotoxic activity in the Ames test.