EMAIL THIS PAGE TO A FRIEND
Archives of pathology & laboratory medicine
Proteomic analysis of oral cavity squamous cell carcinoma specimens identifies patient outcome-associated proteins.
PMID 25295583
Abstract
Global proteomic analysis of oral cavity squamous cell carcinoma was performed to identify changes that reflect patient outcomes. To identify differentially expressed proteins associated with patient outcomes and to explore the use of imaging mass spectrometry as a clinical tool to identify clinically relevant proteins. Two-dimensional separation of digested peptides generated from 43 specimens with high-resolution mass spectrometry identified proteins associated with disease-specific death, distant metastasis, and loco-regional recurrence. RNA expressions had been correlated to protein levels to test transcriptional regulation of clinically relevant proteins. Imaging mass spectrometry explored an alternative platform for assessing clinically relevant proteins that would complement surgical pathologic diagnosis. Seventy-two peptide features were found to be associated with 3 patient outcomes: disease-specific death (9), distant metastasis (16), and loco-regional recurrence (39); 8 of them were associated with multiple outcomes. Functional ontology revealed major changes in cell adhesion and calcium binding. Thirteen RNAs showed strong correlation with their encoded proteins, implying transcriptional control. Reduction of DSP, PKP1, and TRIM29 was associated with significantly shorter time to onset of distant metastasis. Reduction of PKP1 and TRIM29 correlated with poorer disease-specific survival. Additionally, S100A8 and S100A9 reductions were verified for their association with poor prognosis using imaging mass spectrometry, a platform more adaptable for use with surgical pathology. Using global proteomic analysis, we have identified proteins associated with clinical outcomes. The list of clinically relevant proteins observed will provide a means to develop clinical assays for prognosis and optimizing treatment selection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93878
trans-Sinapic acid, analytical standard
C11H12O5