EMAIL THIS PAGE TO A FRIEND
Arquivos de gastroenterologia
Effect of Helicobacter pylori eradication on hepatic steatosis, NAFLD fibrosis score and HSENSI in patients with nonalcoholic steatohepatitis: a MR imaging-based pilot open-label study.
PMID 25296089
Abstract
Limited clinical data suggest Helicobacter pylori (Hp) infection may contribute to nonalcoholic fatty liver disease (NAFLD) pathogenesis. The effect of Hp eradication on hepatic steatosis (magnetic resonance imaging), nonalcoholic fatty liver disease fibrosis score and HSENSI (Homocysteine, serum glutamic oxaloacetic transaminase, Erythrocyte sedimentation rate, nonalcoholic steatohepatitis Index) in nonalcoholic steatohepatitis patients. Thirteen adult patients with biopsy-proven nonalcoholic steatohepatitis, asymptomatic for gastrointestinal disease, underwent 13C urea breath test; Hp positive patients received eradication therapy until repeat test become negative. Hepatic fat fraction, standard biochemical tests and calculation of nonalcoholic fatty liver disease fibrosis score and HSENSI were performed at baseline and month 12. Hepatic fat fraction was similar for between and within group comparisons. Nonalcoholic fatty liver disease fibrosis score showed a non-significant trend towards decrease in Hp(+) [-0.34 (-1.39-0.29) at baseline and -0.24 (-0.99-0.71) at month 12; P = 0.116], whereas increase in Hp(-) group [-0.38 (-1.72-0.11) and -0.56 (-1.43-0.46), respectively; P = 0.249]. HSENSI was significantly decreased only in Hp(+) group [1.0 (1.0-2.0) at baseline and 1.0 (0-1.0) at month 12; P = 0.048]. Hp eradication had no long-term effect on hepatic steatosis, but showed a trend towards improvement in nonalcoholic fatty liver disease fibrosis score and HSENSI. These results warrant larger studies with paired biopsies.