EMAIL THIS PAGE TO A FRIEND
International Microbiology : The Official Journal of the Spanish Society for Microbiology
Relating the outcome of HCV infection and different host SNP polymorphisms in a Majorcan population coinfected with HCV-HIV and treated with pegIFN-RBV.
PMID 25296442