EMAIL THIS PAGE TO A FRIEND
Scientific reports
Self-cleaning Metal Organic Framework (MOF) based ultra filtration membranes--a solution to bio-fouling in membrane separation processes.
PMID 25296745
Abstract
Bio-fouling is a serious problem in many membrane-based separation processes for water and wastewater treatment. Current state of the art methods to overcome this are to modify the membranes with either hydrophilic additives or with an antibacterial compound. In this study, we propose and practise a novel concept to prevent bio-fouling by developing a killing and self-cleaning membrane surface incorporating antibacterial silver nanoparticles and highly hydrophilic negatively charged carboxylic and amine functional groups. The innovative surface chemistry helps to reduce the contact angle of the novel membrane by at least a 48% and increase the pure water flux by 39.4% compared to the control membrane. The flux drop for the novel membrane is also lower (16.3% of the initial flux) than the control membrane (55.3% of the initial flux) during the long term experiments with protein solution. Moreover, the novel membrane continues to exhibit inhibition to microbes even after 1320 min of protein filtration. Synthesis of self-cleaning ultrafiltration membrane with long lasting properties opens up a viable solution for bio-fouling in ultrafiltration application for wastewater purification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31564
Acrylonitrile, analytical standard
C3H3N
110213
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
C3H3N
320137
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
C3H3N
93171
Diethylene glycol, BioUltra, ≥99.0% (GC)
C4H10O3
03128
Diethylene glycol, analytical standard
C4H10O3
32160
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
C4H10O3
PHR1045
Diethylene glycol
C4H10O3
1193265
Diethylene glycol, United States Pharmacopeia (USP) Reference Standard
C4H10O3
H26456
Diethylene glycol, ReagentPlus®, 99%
C4H10O3
63200
Maleic anhydride, puriss., ≥99.0% (NT)
C4H2O3
M188
Maleic anhydride, 99%
C4H2O3
M625
Maleic anhydride, powder, 95%
C4H2O3
ARK2190
N,N-Dimethylacetamide, PharmaGrade, Manufactured under appropriate GMP controls for pharma and biopharmaceutical production., Ph Eur
C4H9NO
72336
N,N-Dimethylacetamide, analytical standard
C4H9NO
270555
N,N-Dimethylacetamide, for HPLC, ≥99.9%
C4H9NO
271012
N,N-Dimethylacetamide, anhydrous, 99.8%
C4H9NO
17308
N,N-Dimethylacetamide, for peptide synthesis, ≥99.8% (GC)
C4H9NO
07-4072
N,N-Dimethylacetamide, SAJ first grade, ≥99.0%
C4H9NO
07-4071
N,N-Dimethylacetamide, SAJ special grade
C4H9NO
185884
N,N-Dimethylacetamide, ReagentPlus®, ≥99%
C4H9NO
522414
N,N-Dimethylacetamide-d9, 99 atom % D
C4D9NO