EMAIL THIS PAGE TO A FRIEND
Journal of bone and mineral metabolism
What is the role of bosentan in healing of femur fractures in a rat model?
PMID 25298328
Abstract
The purpose of this study was to examine the effects bosentan (which is a strong vasoconstrictor) on bone fracture pathophysiology, and investigate the roles of the nonselective endothelinxa01 receptor blocker bosentan on the bone fractures formed in rats through radiographic, histopathologic, and immunohistochemical methods. The rats were divided into three groups (six rats in each group): a femoral fracture control group, a femoral fracturexa0plus bosentan at 50xa0mg/kg group, and a femoral fracture plus bosentan at 100xa0mg/kg group. The femoral fracture model was established by transversely cutting the femur at the midsection. After manual reduction, the fractured femur was fixed with intramedullary Kirschner wires. The radiographic healing scores of the bosentan 100 and 50xa0mg/kg groups were significantly better that those of the fracture control group. The fracture callus percent of new bone in the bosentan 100xa0mg/kg group was significantly greater than that in the control group. Also, semiquantitative analysis showed higher positive vascular endothelial growth factor and osteocalcin staining and lower positive endothelinxa0receptor typexa0A staining in the treatment groups than in the control group. Bosentan treatment also decreased tissue endothelinxa01 expression relative to that in the fracture control group. As a result of our study, the protective effect of bosentan was shown in experimental femoral fracture healing in rats by radiographic, histopathologic, and molecular analyses.