EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Flicker light-induced retinal vasodilation is unaffected by inhibition of epoxyeicosatrienoic acids and prostaglandins in humans.
PMID 25298420
Abstract
To investigate the role of epoxyeicosatrienoic acids (EETs) and prostaglandins (PGs) in retinal blood vessel calibers and vasodilation during flicker light stimulation in humans. Twelve healthy nonsmokers participated in a balanced crossover study. Oral fluconazole 400 mg and dispersible aspirin 600 mg were used to inhibit production of EETs and PGs, respectively. Retinal imaging was performed 1 hour after drug ingestion with the Dynamic Vessel Analyzer. Resting calibers of selected vessel segments were recorded in measurement units (MU). Maximum percentage dilations during flicker stimulation were calculated from baseline calibers. We then studied six participants each after fluconazole and aspirin ingestions at 30-minute intervals for 2 hours. Within-subject differences were assessed by ANOVA and Dunnett-adjusted pairwise comparisons with significance taken at P < 0.05. In crossover study participants, mean (SD) arteriole and venule dilations without drug administration were 4.4% (2.0%) and 4.6% (1.7%), respectively. Neither drug affected vasodilation during flicker stimulation. Mean (SD) resting arteriole and venule calibers on no-drug visits were 119.6 (10.6) MU and 145.7 (17.0) MU, respectively. Fluconazole reduced mean (±95% CI) resting venule calibers by 5.1 (4.3) MU. In repeated measures participants, neither drug affected vasodilations, but fluconazole reduced resting venule calibers over 2 hours (P < 0.001). Epoxyeicosatrienoic acids and prostaglandins are unlikely to be primary mediators of flicker light-induced retinal vasodilation in humans. However, EETs may play a role in the regulation of retinal vascular tone and blood flow under resting physiological conditions.