EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Some comments on the type references of the official nomenclature (IUB) for beta-N-acetylglucosaminidase, beta-N-acetylhexosaminidase and beta-N-acetylgalactosaminidase.
PMID 2529847