EMAIL THIS PAGE TO A FRIEND
Bioconjugate chemistry
Cell penetrating peptide (CPP)-conjugated desferrioxamine for enhanced neuroprotection: synthesis and in vitro evaluation.
PMID 25299707
Abstract
Iron overload causes progressive and sometimes irreversible damage due to accelerated production of reactive oxygen species. Desferrioxamine (DFO), a siderophore, has been used clinically to remove excess iron. However, the applications of DFO are limited because of its inability to access intracellular labile iron. Cell penetrating peptides (CPPs) have become an efficient delivery vector for the enhanced internalization of drugs into the cytosol. We describe, herein, an efficient method for covalently conjugating DFO to the CPPs TAT(47-57) and Penetratin. Both conjugates suppressed the redox activity of labile plasma iron in buffered solutions and in iron-overloaded sera. Enhanced access to intracellular labile iron compared to the parent siderophore was achieved in HeLa and RBE4 (a model of blood-brain-barrier) cell lines. Iron complexes of both conjugates also had better permeability in both cell models. DFO antioxidant and iron binding properties were preserved and its bioavailability was increased upon CPP conjugation, which opens new therapeutic possibilities for neurodegenerative processes associated with brain iron overload.