EMAIL THIS PAGE TO A FRIEND
Environmental Science and Pollution Research International
Impact of soil primary size fractions on sorption and desorption of atrazine on organo-mineral fractions.
PMID 25300187