EMAIL THIS PAGE TO A FRIEND
Eksperimentalna meditsina i morfologiia
[Study of the effect of analgeton on the release of beta-endorphin and its effect on chronic hyperalgesia].
PMID 2530083
Abstract
The effect of the combined preparation analgeton and its components--analgin (metamizol, noramidopyrindimethansulfonate) and aminton (2-amino-4-methyl-pyridinphosphate) was studied on the release of beta-endorphin and its influence on chronic hyperalgesia. Experiments were carried out in vivo and in vitro on white rats and it was established that analgin, aminton and analgeton stimulated the release of beta-endorphin, affecting various regulatory levels. Analgeton in contrast to analgin showed longer analgetic effect in animals with chronic hyperalgesia (adjuvant arthritis of white rats). Possibilities for pharmacological control of chronic pain by influencing the levels of endogenous opioids are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123080
2-Amino-4-methylpyridine, 99%
C6H8N2