EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology: Gastrointestinal and Liver Physiology
Mesenchymal stem cells and conditioned medium avert enteric neuropathy and colon dysfunction in guinea pig TNBS-induced colitis.
PMID 25301186