EMAIL THIS PAGE TO A FRIEND
Cancer Letters
SKLB316, a novel small-molecule inhibitor of cell-cycle progression, induces G2/M phase arrest and apoptosis in vitro and inhibits tumor growth in vivo.
PMID 25301449