EMAIL THIS PAGE TO A FRIEND
Journal of medical microbiology
Characterization of Streptococcus pneumoniae clones from paediatric patients with cystic fibrosis.
PMID 25301526
Abstract
The role of Streptococcus pneumoniae in cystic fibrosis (CF) is poorly understood. The pneumococcal population has changed over time after the introduction of the heptavalent conjugate vaccine (PCV7) and, more recently, the 13-valent conjugate vaccine (PCV13). Although serotypes and clones causing invasive pneumococcal disease or colonizing healthy children have been extensively analysed, little is known so far on the serotypes and clones of pneumococci in CF patients. The aim of this work was to investigate serotypes, antibiotic susceptibilities, genotypes and biofilm production of CF pneumococcal isolates. Overall, 44 S. pneumoniae strains collected from 32 paediatric CF patients from January 2010 to May 2012 in a large Italian CF Centre were tested for antimicrobial susceptibility testing by Etest, serotyped by the Quellung reaction and genotyped by a combination of different molecular typing methods, including pbp gene restriction profiling, pspA restriction profiling and sequencing, PFGE and multilocus sequence typing. Biofilm production by pneumococcal strains was also assessed. Penicillin non-susceptibility was 16 %. High resistance rates (>56 %) were observed for erythromycin, clindamycin and tetracycline. The most frequent serotype recovered was serotype 3 (31.8 %). The coverage of PCV7 and PCV13 was 6.8 and 47.7 %, respectively. More than 80 % of CF strains belonged to Pneumococcal Molecular Epidemiology Network (PMEN) reference clones, the most common being Netherlands(3)-ST180 (28.2 %), and Greece(21)-30/ST193 (15.4 %). All strains produced biofilm in vitro, although with large variability in biofilm formation efficiency. No correlation was found between biofilm levels and serotype, clone or antibiotic resistance. The high isolation rate of antibiotic-resistant serotype 3 pneumococci from CF patients suggests that PCV13 could increase protection from pneumococcal colonization and infection.