EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Mechanistic insight into the formation of cationic naked nanocrystals generated under equilibrium control.
PMID 25302526
Abstract
Cationic naked nanocrystals (NCs) are useful building units for assembling hierarchical mesostructured materials. Until now, their preparation required strongly electrophilic reagents that irreversibly sever bonds between native organic ligands and the NC surface. Colloidal instabilities can occur during ligand stripping if exposed metal cations desorb from the surface. We hypothesized that cation desorption could be avoided were we able to stabilize the surface during ligand stripping via ion pairing. We were successful in this regard by carrying out ligand stripping under equilibrium control with Lewis acid-base adducts of BF3. To better understand the microscopic processes involved, we studied the reaction pathway in detail using in situ NMR experiments and electrospray ionization mass spectrometry. As predicted, we found that cationic NC surfaces are transiently stabilized post-stripping by physisorbed anionic species that arise from the reaction of BF3 with native ligands. This stabilization allows polar dispersants to reach the NC surface before cation desorption can occur. The mechanistic insights gained in this work provide a much-needed framework for understanding the interplay between NC surface chemistry and colloidal stability. These insights enabled the preparation of stable naked NC inks of desorption-susceptible NC compositions such as PbSe, which were easily assembled into new mesostructured films and polymer-nanocrystal composites with wide-ranging technological applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
296309
1,4-Dioxane, anhydrous, 99.8%
C4H8O2
533971
1,4-Dioxane, histological grade, ≥99%
C4H8O2
76887
1,4-Dioxane, analytical standard
C4H8O2
07-5360
1,4-Dioxane, SAJ first grade
C4H8O2
07-5390
1,4-Dioxane, for HPLC
C4H8O2
PHR1549
1,4-Dioxane, Pharmaceutical Secondary Standard; Certified Reference Material
C4H8O2
74738
1-Octadecene, analytical standard
C18H36
74740
1-Octadecene, ≥95.0% (GC)
C18H36
74745
1-Octadecene, technical, ~85% (GC)
C18H36
463086
Boron trifluoride, electronic grade, ≥99.99%
BF3
295027
Boron trifluoride, ≥99.5%
BF3
252964
Diphenylphosphine, 98%
C12H11P
52730
Hexamethylphosphoramide, purum, ≥98.0% (GC)
C6H18N3OP
779903
Hexamethylphosphoramide, absolute, over molecular sieve (H2O ≤0.01%), ≥98.0% (GC)
C6H18N3OP
H11602
Hexamethylphosphoramide, 99%
C6H18N3OP
189979
N,N-Dimethylformamide-d7, ≥99.5 atom % D
C3D7NO
700428
N,N-Dimethylformamide-d7, ≥99.5 atom % D, contains 0.03 % (v/v) TMS
C3D7NO
269905
N,N-Dimethylformamide-d7, ≥99.5 atom % D, contains 1 % (v/v) TMS
C3D7NO
296988
Octane, anhydrous, ≥99%
C8H18
657042
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
C8H18
74820
Octane, analytical standard
C8H18
117854
Trioctylphosphine, technical grade, 90%
C24H51P
92840
Trioctylphosphine, technical, ~90% (GC)
C24H51P
718165
Trioctylphosphine, 97%
C24H51P
442272
1-Octadecene, analytical standard
C18H36