EMAIL THIS PAGE TO A FRIEND
Clinical and Experimental Immunology
Depletion of regulatory T cells in a hapten-induced inflammation model results in prolonged and increased inflammation driven by T cells.
PMID 25302741