PloS one
Increased activity of cell membrane-associated prothrombinase, fibrinogen-like protein 2, in peripheral blood mononuclear cells of B-cell lymphoma patients.
PMID 25303152
Abstract
Fibrinogen-like protein 2, FGL-2, was reported to be overexpressed in various cancer tissues, where it acts as a transmembrane prothrombinase. This study aims to determine the prothrombinase activity of FGL-2 in peripheral blood mononuclear cells (PBMC) of patients with B-cell lymphoma. FGL-2 activity was determined in patients with B-cell lymphoma (n = 53), and healthy controls (n = 145). FGL-2 activity in patients at diagnosis increased 3 ± 0.3 fold (p < 0.001). Sensitivity and specificity of the test was established at 73.6% and 80.7%, respectively, using a cutoff of 150% activity over control. Moreover, FGL-2 activity in 10 of 11 patients in remission decreased by 76%. In contrast, no significant difference was observed in expression levels of fgl-2 gene in patients and controls. Taken together, our study indicates that FGL-2 prothrombinase activity in PBMC of lymphoma patients is increased in active disease and normalizes during remission, thus being a potential marker for follow up of lymphoma patients.