EMAIL THIS PAGE TO A FRIEND
Bone
Whole body vibration improves osseointegration by up-regulating osteoblastic activity but down-regulating osteoblast-mediated osteoclastogenesis via ERK1/2 pathway.
PMID 25304090