EMAIL THIS PAGE TO A FRIEND
Experimental Biology and Medicine (Maywood, N.J.)
No evidence of association between variant rs2075650 in lipid metabolism-related locus APOE/TOMM40 and advanced age-related macular degeneration in Han Chinese population.
PMID 25304313