EMAIL THIS PAGE TO A FRIEND
Toxicology and Applied Pharmacology
Protective effect of nuclear factor E2-related factor 2 on inflammatory cytokine response to brominated diphenyl ether-47 in the HTR-8/SVneo human first trimester extravillous trophoblast cell line.
PMID 25305463
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
112941
tert-Butylhydroquinone, 97%
C10H14O2