EMAIL THIS PAGE TO A FRIEND
Acta Biomaterialia
Hyaluronic acid-tumor necrosis factor-related apoptosis-inducing ligand conjugate for targeted treatment of liver fibrosis.
PMID 25305513
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
71859
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
INaO4
B91005
tert-Butyl carbazate, 98%
C5H12N2O2