EMAIL THIS PAGE TO A FRIEND
The Science of the Total Environment
Identification of Triclosan-O-Sulfate and other transformation products of Triclosan formed by activated sludge.
PMID 25306094
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35811
2,4-Dichlorophenol, PESTANAL®, analytical standard
C6H4Cl2O
48528
2,4-Dichlorophenol, analytical standard
C6H4Cl2O
105953
2,4-Dichlorophenol, 99%
C6H4Cl2O
36130
2,4-Dichlorophenol, purum, ≥97.0% (HPLC)
C6H4Cl2O
07846
2,4-Dichlorophenol, certified reference material, TraceCERT®
C6H4Cl2O
148520
4-Chlorocatechol, 97%
C6H5ClO2
524190
5-Chloro-2-(2,4-dichlorophenoxy)phenol, 97%
C12H7Cl3O2
72779
Irgasan, ≥97.0% (HPLC)
C12H7Cl3O2
194166
Methanol-d4, "100%", ≥99.96 atom % D, ≥99% (CP)
CD4O
721948 Methanol-d4, NMR reference standard, 99.8 atom % D, NMR tube size 5 mm × 8 in.
CD4O
721921 Methanol-d4, NMR reference standard, 99.8 atom % D, NMR tube size 3 mm × 8 in.
CD4O
444758
Methanol-d4, "100%", 99.96 atom % D
CD4O
423084
Methanol-d4, "100%", 99.96 atom % D
CD4O
535435
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
CD4O
151947
Methanol-d4, ≥99.8 atom % D
CD4O
522023
Methanol-d4, 99.8 atom % D
CD4O
441384
Methanol-d4, ≥99.8 atom % D
CD4O
343803
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
CD4O
530530
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
CD4O
611646
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
CD4O
439029
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
CD4O
417653
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
CD4O
422878
Methanol-d4, 99 atom % D
CD4O
569534
Methanol-d4, anhydrous, ≥99.8 atom % D
CD4O
570729
Methanol-d4, ≥99.8 atom % D, anhydrous
CD4O
87920
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
C4H12Si
87921
Tetramethylsilane, ≥99.0% (GC)
C4H12Si
744301
Tetramethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
C4H12Si
523771
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
C4H12Si
T24007
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
C4H12Si