EMAIL THIS PAGE TO A FRIEND
Molecular Genetics and Metabolism
Phenotypic heterogeneity in monogenic diabetes: the clinical and diagnostic utility of a gene panel-based next-generation sequencing approach.
PMID 25306193