EMAIL THIS PAGE TO A FRIEND
Nature methods
A DNA-based molecular probe for optically reporting cellular traction forces.
PMID 25306545
Abstract
We developed molecular tension probes (TPs) that report traction forces of adherent cells with high spatial resolution, can in principle be linked to virtually any surface, and obviate monitoring deformations of elastic substrates. TPs consist of DNA hairpins conjugated to fluorophore-quencher pairs that unfold and fluoresce when subjected to specific forces. We applied TPs to reveal that cellular traction forces are heterogeneous within focal adhesions and localized at their distal edges.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R4500
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
C17H20N4O6
R7649
(−)-Riboflavin, meets USP testing specifications
C17H20N4O6
R9504
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
C17H20N4O6
86760
1,1,2,2-Tetrabromoethane, purum p.a., for separation (of mineral compounds), ≥98.0% (GC)
C2H2Br4
185574
1,1,2,2-Tetrabromoethane, 98%
C2H2Br4
588857
3-(Ethoxydimethylsilyl)propylamine, 97%
C7H19NOSi
92846
5-Carboxyfluorescein N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
C25H15NO9
295000
Argon, ≥99.998%
Ar
S4250
D-Serine, ≥98% (TLC)
C3H7NO3
613339
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
CH3NO
551384
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO
612707
Formamide solution, NMR reference standard, 45% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
CH3NO
611891
Formamide solution, NMR reference standard, 45% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO
1365000
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
C15H11I4NO4
56480
N-Hydroxysuccinimide, purum, ≥97.0% (T)
C4H5NO3
77608
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
C6H6O
35952
Phenol, PESTANAL®, analytical standard
C6H6O
PHR1047
Phenol
C6H6O
1524806
Phenol, United States Pharmacopeia (USP) Reference Standard
C6H6O
46344
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C6H6O
240958
Propylamine, ≥99%
C3H9N
109819
Propylamine, 98%
C3H9N
53646
Propylamine, analytical standard
C3H9N
82100
Propylamine, purum, ≥99.0% (GC)
C3H9N
PHR1054
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
C17H20N4O6
1603006
Riboflavin, United States Pharmacopeia (USP) Reference Standard
C17H20N4O6
R0600000
Riboflavin, European Pharmacopoeia (EP) Reference Standard
C17H20N4O6
47861
Riboflavin (B2), analytical standard
C17H20N4O6
Y0000757
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
C17H20N4O6
IRMM468
Thyroxine (T4), IRMM® certified Reference Material
C15H11I4NO4