EMAIL THIS PAGE TO A FRIEND
Biochimica et Biophysica Acta
The peculiar N- and (-termini of trichogin GA IV are needed for membrane interaction and human cell death induction at doses lacking antibiotic activity.
PMID 25306964