EMAIL THIS PAGE TO A FRIEND
Pesticide biochemistry and physiology
Acute toxicity and sublethal effects of fipronil on detoxification enzymes in juvenile zebrafish (Danio rerio).
PMID 25307460
Abstract
The acute toxicity of fipronil and its sublethal effects on detoxification enzymes (carboxylesterases (CarEs), glutathione S-transferases (GSTs), and 7-ethoxycoumarin O-deethylase (ECOD)) in zebrafish (Danio rerio) were investigated. The results indicated that the 24-h LC50 of fipronil for zebrafish was 220.4 μg/L (95% CI: 173.7-272.4 μg/L). Sublethal concentrations of fipronil did not cause significant changes in CarEs activities. In the liver and muscle tissues, GST activities at the tested concentrations did not significantly differ from those in the control. In the brain and gill tissues, GST activities at a concentration of 4 μg/L were significantly lower than those at a concentration of 2 μg/L. The results suggest that CarEs and GSTs were not suitable biomarkers for fipronil effects in D. rerio. A significant induction in the ECOD activities in the brain, gill, liver, and muscle tissues was observed compared with the control. Moreover, the dose-dependent responses of the ECOD activity were observed after treatment with sublethal concentrations of fipronil in the range of 2-20 μg/L. The results suggested that ECOD could be a suitable biomarker of fipronil effects in D. rerio.