EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
New insights into the peroxisomal protein inventory: Acyl-CoA oxidases and -dehydrogenases are an ancient feature of peroxisomes.
PMID 25307522
Abstract
Peroxisomes are ubiquitous organelles which participate in a variety of essential biochemical pathways. An intimate interrelationship between peroxisomes and mitochondria is emerging in mammals, where both organelles cooperate in fatty acid β-oxidation and cellular lipid homeostasis. As mitochondrial fatty acid β-oxidation is lacking in yeast and plants, suitable genetically accessible model systems to study this interrelationship are scarce. Here, we propose the filamentous fungus Ustilago maydis as a suitable model for those studies. We combined molecular cell biology, bioinformatics and phylogenetic analyses and provide the first comprehensive inventory of U. maydis peroxisomal proteins and pathways. Studies with a peroxisome-deficient Δpex3 mutant revealed the existence of parallel and complex, cooperative β-oxidation pathways in peroxisomes and mitochondria, mimicking the situation in mammals. Furthermore, we provide evidence that acyl-CoA dehydrogenases (ACADs) are bona fide peroxisomal proteins in fungi and mammals and together with acyl-CoA oxidases (ACOX) belong to the basic enzymatic repertoire of peroxisomes. A genome comparison with baker's yeast and human gained new insights into the basic peroxisomal protein inventory shared by humans and fungi and revealed novel peroxisomal proteins and functions in U. maydis. The importance of our findings for the evolution and function of the complex interrelationship between peroxisomes and mitochondria in fatty acid β-oxidation is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19215
Butyric acid, analytical standard
C4H8O2
B103500
Butyric acid, ≥99%
C4H8O2
W222100
Butyric acid, ≥99%, FG
C4H8O2
W222119
Butyric acid, natural, ≥99%, FCC, FG
C4H8O2
W236403
Decanoic acid, ≥99.5%, FCC, FG
C10H20O2
W236411
Decanoic acid, natural, ≥98%, FCC, FG
C10H20O2
C1875
Decanoic acid, ≥98.0%
C10H20O2
21409
Decanoic acid, analytical standard
C10H20O2
E3385
Erucic acid, ≥99% (capillary GC)
C22H42O2
45629
Erucic acid, analytical standard
C22H42O2
45630
Erucic acid, technical, ~90% (GC)
C22H42O2
153745
Hexanoic acid, ≥99%
C6H12O2
W255904
Hexanoic acid, ≥98%, FCC, FG
C6H12O2
21529
Hexanoic acid, analytical standard
C6H12O2
21530
Hexanoic acid, purum, ≥98.0% (GC)
C6H12O2
L6641
Lignoceric acid, ≥99% (GC)
C24H48O2
M3128
Myristic acid, Sigma Grade, ≥99%
C14H28O2
70079
Myristic acid, analytical standard
C14H28O2
70082
Myristic acid, ≥98.0% (GC)
C14H28O2
W276405
Myristic acid, ≥95%, FCC, FG
C14H28O2
W276413
Myristic acid, natural, ≥98.5%, FG
C14H28O2
130087
Myristic acid, 95%
C14H28O2
PHR1124
Myristic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C14H28O2
41788
Myristoleic acid, analytical standard
C14H26O2
M3525
Myristoleic acid, ≥99% (capillary GC)
C14H26O2
75090
Oleic acid, analytical standard
C18H34O2
Y0001479
Oleic acid, European Pharmacopoeia (EP) Reference Standard
C18H34O2
05508
Oleic acid, Selectophore, ≥99.0%
C18H34O2
P0500
Palmitic acid, ≥99%
C16H32O2
43051
Palmitic acid, certified reference material, TraceCERT®
C16H32O2