EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Long-term storage of lyophilized liposomal formulations.
PMID 25308534
Abstract
Because aqueous liposomal formulations containing multiply unsaturated lipids are susceptible to chemical degradation, these formulations are often lyophilized. Despite their limited chemical stability, interest in the use of multiply unsaturated lipids to promote intracellular delivery has increased considerably in recent years. The goal of the current study was to examine the long-term storage stability of lyophilized formulations containing lipids with increasing levels of unsaturation, and various strategies that can be employed to improve stability. Aqueous lipid-trehalose formulations containing 1,2-dilinolenoyl-sn-glycero-3-phosphocholine (DLPC), 1,2-dilinoleoyl-sn-glycero-3-phosphocholine (DLinPC), or 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC) were lyophilized and stored at temperatures ranging from 4°C to 60°C. We observed that the lipid degradation rate increased as the storage temperature and unsaturation level were increased. Even the cleanest sugars, which are available commercially, contain iron contaminants, and it was observed that the chelation of these iron contaminants significantly improved the stability of DLPC during storage. However, the glass transition temperature of the sugar that was included in the formulation, the reduction of the oxygen in the aqueous sample prior to lyophilization, the inclusion of helper lipids (i.e., cholesterol), and the rate of freezing did not significantly improve stability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90872
Acetone, analytical standard
C3H6O
PHR1219
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
C3H6O
1006801
Acetone, United States Pharmacopeia (USP) Reference Standard
C3H6O
295000
Argon, ≥99.998%
Ar
02487
Chloroform, analytical standard
CHCl3
PHR1552
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
CHCl3
S1876
D-Sorbitol, ≥98%
C6H14O6
85529
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
C6H14O6
S6021
D-Sorbitol, for molecular biology, ≥98%
C6H14O6
240850
D-Sorbitol, 99%
C6H14O6
S3889
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
C6H14O6
97336
D-Sorbitol, liquid, tested according to Ph Eur
C6H14O6
47841
D-Sorbitol, analytical standard
C6H14O6
S7547
D-Sorbitol, BioXtra, ≥98%
C6H14O6
W302902
D-Sorbitol, FCC, FG
C6H14O6
PHR1553
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO
17933
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
HCl
17934
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
HCl
17935
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
HCl
426466
Methylamine solution, 40 wt. % in H2O
CH5N
534102
Methylamine solution, 33 wt. % in absolute ethanol
CH5N
395056
Methylamine solution, 2.0 M in THF
CH5N
395048
Methylamine solution, 2.0 M in methanol
CH5N
D4551
N,N-Dimethylformamide, for molecular biology, ≥99%
C3H7NO
72438
N,N-Dimethylformamide, analytical standard
C3H7NO
402907
Propionic acid, ACS reagent, ≥99.5%
C3H6O2
94425
Propionic acid, analytical standard
C3H6O2
W292400
Propionic acid, ≥99.5%, FCC, FG
C3H6O2
81910
Propionic acid, puriss. p.a., ≥99.5% (GC)
C3H6O2
P5561
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
C3H6O2