EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Comparison of point-of-care tests for the rapid diagnosis of visceral leishmaniasis in East African patients.
PMID 25311696
Abstract
The development of rK39-based rapid diagnostic tests (RDTs) has greatly aided the diagnosis of visceral leishmaniasis, especially in the Indian subcontinent and Brazil, by offering high sensitivity and specificity. However, these tests have been less sensitive and less specific in sub-Saharan Africa. To improve upon the performance of rK39 in Africa, we engineered the fusion molecule rK28, which retained some of the rK39 repeats and combined them with repeat sequences from two additional Leishmania genes. This polyprotein was used in the development of several prototype RDTs by different commercial manufacturers with the goal of assessing relative performance in inexpensive formats. Here, we report field studies showing that the rK28 antigen could be readily adapted to a variety of RDT formats to achieve high sensitivity, generally > 90%, and adequate specificity to aid in the diagnosis of human visceral leishmaniasis in East Africa, Asia, and South America.