EMAIL THIS PAGE TO A FRIEND
Infection and Immunity
Macrophages are the determinant of resistance to and outcome of nonlethal Babesia microti infection in mice.
PMID 25312951
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4434
Dichloromethylenediphosphonic acid disodium salt
CH2Cl2Na2O6P2