EMAIL THIS PAGE TO A FRIEND
Saudi medical journal
Clostridium difficile in retail baskets, trolleys, conveyor belts, and plastic bags in Saudi Arabia.
PMID 25316477
Abstract
To determine Clostridium difficile (C. difficile) prevalence on retail surfaces and shoppers plastic bags. From 20 June to 10 August 2011, in a cross-sectional epidemiological study, 17 supermarkets from 2 cities, Albaha and Altaif, Saudi Arabia were sampled. A total of 800 samples, which comprised 200 samples per surveyed surface, were studied. These included baskets, trolleys, conveyer belts, and outgoing shoppers' plastic bags. Clostridium difficile strains were isolated. The isolates were characterized using ribotyping and  polymerase chain reaction for the detection of toxin A (tcdA), toxin B (tcdB), binary toxin (cdtB), and toxin C (tcdC) genes. Susceptibility to antibiotics was determined on a Muller-Hinton agar with 5% sheep blood agar using E-tests. Overall, the C. difficile prevalence on sampled surfaces was 0.75%. The highest prevalence was found on retail baskets and trolleys, followed by plastic bags. A total of 5 different ribotypes were identified. Alterations in tcdC were detected in ribotype 027 and BT1. All the identified isolates were susceptible to vancomycin, but resistant to levofloxacin. In this study, C. difficile was present at a rate of 0.75% on supermarket surfaces. Spore disinfection of implicated surfaces may be necessary to control any community-acquired infections caused by this pathogen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31140
L-(+)-Fructose, ≥97.0% (HPLC)
C6H12O6
40922
Levofloxacin, analytical standard
C18H20FN3O4