EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Medicine
The marine-derived fungal metabolite, terrein, inhibits cell proliferation and induces cell cycle arrest in human ovarian cancer cells.
PMID 25318762