EMAIL THIS PAGE TO A FRIEND
Chemical Biology & Drug Design
4-Substituted-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine derivatives: design, synthesis, antitumor and EGFR tyrosine kinase inhibitory activity.
PMID 25318985