EMAIL THIS PAGE TO A FRIEND
International Journal of Cosmetic Science
Preliminary 1 month stability screening of cosmetic multiple emulsions (W/O/W) prepared using cetyl dimethicone copolyol and Polysorbate 80.
PMID 25319627