EMAIL THIS PAGE TO A FRIEND
ChemMedChem
[(3) H]UR-DE257: development of a tritium-labeled squaramide-type selective histamine H2 receptor antagonist.
PMID 25320025
Abstract
A series of new piperidinomethylphenoxypropylamine-type histamine H2 receptor (H2 R) antagonists with different substituted "urea equivalents" was synthesized and characterized in functional in vitro assays. Based on these data as selection criteria, radiosynthesis of N-[6-(3,4-dioxo-2-{3-[3-(piperidin-1-ylmethyl)phenoxy]propylamino}cyclobut-1-enylamino)hexyl]-(2,3-(3) H2 )propionic amide ([(3) H]UR-DE257) was performed. The radioligand (specific activity: 63 Ci mmol(-1) ) had high affinity for human, rat, and guinea pig H2 R (hH2 R, Sf9 cells: Kd , saturation binding: 31 nM, kinetic studies: 20 nM). UR-DE257 revealed high H2 R selectivity on membranes of Sf9 cells, expressing the respective hHx R subtype (Ki values: hH1 R: >10000 nM, hH2 R: 28 nM, hH3 R: 3800 nM, hH4 R: >10000 nM). In spite of insurmountable antagonism, probably due to rebinding of [(3) H]UR-DE257 to the H2 R (extended residence time), the title compound proved to be a valuable pharmacological tool for the determination of H2 R affinities in competition binding assays.