EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology
Fructose- and glucose-conditioned preferences in FVB mice: strain differences in post-oral sugar appetition.
PMID 25320345