EMAIL THIS PAGE TO A FRIEND
Cell Death & Disease
Medicarpin, a legume phytoalexin sensitizes myeloid leukemia cells to TRAIL-induced apoptosis through the induction of DR5 and activation of the ROS-JNK-CHOP pathway.
PMID 25321472
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU061431 MISSION® esiRNA, esiRNA human TNFRSF10B (esiRNA1)