EMAIL THIS PAGE TO A FRIEND
Molecular Medicine Reports
Short hairpin RNA targeting Notch2 inhibits U87 human glioma cell proliferation by inducing cell cycle arrest and apoptosis in vitro and in vivo.
PMID 25323114